Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromothiophene-2-methanol 98%

Supplier: AMBEED, INC
Warning

A116921-10G A116921-1G A116921-25G
77136-322EA 37.08 USD InStock
77136-322 77136-324 77136-326
4-Bromothiophene-2-methanol 98%
4-Bromothiophene-2-methanol


Formula: C₅H₅BrOS
MDL Number: MFCD04115392
CAS Number: 79757-77-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty