Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Naphthaldehyde 95%

Supplier: AMBEED, INC
Warning

A117528-1000G A117528-100G A117528-10G A117528-25G A117528-500G
77136-340EA 275.01 USD InStock
77136-340 77136-342 77136-344 77136-346 77136-348
1-Naphthaldehyde 95%
1-Naphthaldehyde
Precaución: for Research Use Only


Formula: C₁₁H₈O
MW: 156.18 g/mol
Boiling Pt: 292
Melting Pt: 2
MDL Number: MFCD00004003
CAS Number: 66-77-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty