Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[(4-Chlorobenzoyl)amino]acetic acid 95%

Supplier: AMBEED, INC
Warning

A119254-1G A119254-250MG A119254-5G
77136-426EA 63.86 USD InStock
77136-426 77136-428 77136-430
2-[(4-Chlorobenzoyl)amino]acetic acid 95%
2-[(4-Chlorobenzoyl)amino]acetic acid
Precaución: for Research Use Only


Formula: C₉H₈ClNO₃
MDL Number: MFCD00020447
CAS Number: 13450-77-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty