Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Fluoro-3-isopropoxy-5-methylphenyl)boronic acid 97%

Supplier: AMBEED, INC
Warning

A124037-1G A124037-250MG A124037-5G
77136-630EA 55.62 USD InStock
77136-630 77136-632 77136-634
(2-Fluoro-3-isopropoxy-5-methylphenyl)boronic acid 97%
(2-Fluoro-3-isopropoxy-5-methylphenyl)boronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₄BFO₃
MW: 212.02 g/mol
MDL Number: MFCD19981515
CAS Number: 2096336-18-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty