Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethoxy(3-glycidyloxypropyl)methylsilane 98%

Supplier: AMBEED, INC
Warning

A125655-1000G A125655-25G A125655-500G
77136-708EA 132.87 USD InStock
77136-708 77136-710 77136-712
Diethoxy(3-glycidyloxypropyl)methylsilane 98%
Diethoxy(3-glycidyloxypropyl)methylsilane
Precaución: for Research Use Only


Formula: C₁₁H₂₄O₄Si
MDL Number: MFCD00046995
CAS Number: 2897-60-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty