Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-tert-Butyl 3a,4,7,7a-tetrahydro-1H-isoindole-2(3H)-carboxylate 97%

Supplier: AMBEED, INC
Warning

A130143-100MG A130143-1G A130143-250MG
77136-886EA 7.21 USD InStock
77136-886 77136-888 77136-890
cis-tert-Butyl 3a,4,7,7a-tetrahydro-1H-isoindole-2(3H)-carboxylate 97%
cis-tert-Butyl 3a,4,7,7a-tetrahydro-1H-isoindole-2(3H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₂₁NO₂
MDL Number: MFCD23378545
CAS Number: 474925-37-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty