Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Difluoro-2-methyl-5-nitrobenzene 97%

Supplier: AMBEED, INC
Warning

A130408-10G A130408-1G A130408-250MG A130408-25G
77136-894EA 23.69 USD InStock
77136-894 77136-896 77136-898 77136-900
1,4-Difluoro-2-methyl-5-nitrobenzene 97%
1,4-Difluoro-2-methyl-5-nitrobenzene
Precaución: for Research Use Only


Formula: C₇H₅F₂NO₂
MDL Number: MFCD06200905
CAS Number: 141412-60-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty