Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Hydroxynaphthalimide trifluoromethanesulfonate 97%

Supplier: AMBEED, INC
Danger

A130840-1G A130840-5G
77136-906EA 13.39 USD InStock
77136-906 77136-908
N-Hydroxynaphthalimide trifluoromethanesulfonate 97%
N-Hydroxynaphthalimide trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₁₃H₆F₃NO₅S
MDL Number: MFCD02683478
CAS Number: 85342-62-7
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty