Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)isoindoline-1,3-dione 98%

Supplier: AMBEED, INC
Warning

A132527-10G A132527-1G A132527-250MG A132527-5G
77136-952EA 190.56 USD InStock
77136-952 77136-954 77136-956 77136-958
2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)isoindoline-1,3-dione 98%
2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)isoindoline-1,3-dione
Precaución: for Research Use Only


Formula: C₂₁H₂₂BNO₄
MDL Number: MFCD02179490
CAS Number: 214360-75-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty