Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-iodotoluene 95%

Supplier: AMBEED, INC
Warning

A133434-100G A133434-10G A133434-25G A133434-500G
77136-984EA 59.74 USD InStock
77136-984 77136-986 77136-988 77136-990
4-Chloro-2-iodotoluene 95%
4-Chloro-2-iodotoluene
Precaución: for Research Use Only


Formula: C₇H₆ClI
MW: 252.48
MDL Number: MFCD00060661
CAS Number: 33184-48-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty