Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-(trifluoromethyl)aniline 98%

Supplier: AMBEED, INC
Warning

A134302-100G A134302-10G A134302-1G A134302-25G A134302-500G A134302-5G
77137-006EA 39.14 USD InStock
77137-006 77137-008 77137-010 77137-012 77137-014 77137-016
3-Bromo-5-(trifluoromethyl)aniline 98%
3-Bromo-5-(trifluoromethyl)aniline
Precaución: for Research Use Only


Formula: C₇H₅BrF₃N
MW: 240.02
MDL Number: MFCD00236205
CAS Number: 54962-75-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty