Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-nitrobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A136329-1000G A136329-100G A136329-25G A136329-500G A136329-5G
77137-096EA 80.34 USD InStock
77137-096 77137-098 77137-100 77137-102 77137-104
4-Chloro-2-nitrobenzoic acid 97%
4-Chloro-2-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄ClNO₄
MDL Number: MFCD00007214
CAS Number: 6280-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty