Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-4-iodobenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A139730-1G A139730-250MG A139730-5G A139730-25G
77137-250EA 23.69 USD InStock
77137-250 77137-252 77137-254 77685-195
3-Hydroxy-4-iodobenzaldehyde 98%
3-Hydroxy-4-iodobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₅IO₂
MDL Number: MFCD00114697
CAS Number: 135242-71-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty