Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Bromophenyl)-2-thiourea 98%

Supplier: AMBEED, INC
Danger

A140703-25G A140703-5G
77137-286EA 54.59 USD InStock
77137-286 77137-288
1-(4-Bromophenyl)-2-thiourea 98%
1-(4-Bromophenyl)-2-thiourea
Precaución: for Research Use Only


Formula: (C₆H₄Br)NHC(S)NH₂
MDL Number: MFCD00041176
CAS Number: 2646-30-2
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty