Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Difluoromethoxy)-6-fluorobenzylamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A141312-100MG A141312-1G A141312-250MG
77137-322EA 7.21 USD InStock
77137-322 77137-324 77137-326
2-(Difluoromethoxy)-6-fluorobenzylamine hydrochloride 95%
2-(Difluoromethoxy)-6-fluorobenzylamine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₉ClF₃NO
MDL Number: MFCD20731225
CAS Number: 1354951-46-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty