Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-chlorobenzoxazole 97%

Supplier: AMBEED, INC
Warning

A144238-10G A144238-10MG A144238-1G A144238-50MG A144238-5G A144238-100G
77137-460EA 100.94 USD InStock
77137-460 77137-462 77137-464 77137-466 77137-468 77685-224
2-Amino-5-chlorobenzoxazole 97%
2-Amino-5-chlorobenzoxazole
Precaución: for Research Use Only


Formula: C₇H₅ClN₂O
MW: 168.58
Boiling Pt: 309
Melting Pt: 184
MDL Number: MFCD00005770
CAS Number: 61-80-3
Merck Index: 12,10328
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty