Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-iodopyridine 97%

Supplier: AMBEED, INC
Warning

A152736-1G A152736-25G A152736-5G A152736-10G
77137-838EA 13.39 USD InStock
77137-838 77137-840 77137-842 77685-263
3-Chloro-4-iodopyridine 97%
3-Chloro-4-iodopyridine
Precaución: for Research Use Only


Formula: C₅H₃ClIN
MW: 239.44 g/mol
MDL Number: MFCD00272199
CAS Number: 77332-79-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty