Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-nitro-p-toluidine 97%

Supplier: AMBEED, INC
Warning

A152987-10G A152987-1G A152987-25G A152987-5G
77137-854EA 33.99 USD InStock
77137-854 77137-856 77137-858 77137-860
5-Chloro-2-nitro-p-toluidine 97%
5-Chloro-2-nitro-p-toluidine
Precaución: for Research Use Only


Formula: C₇H₇ClN₂O₂
MDL Number: MFCD00034066
CAS Number: 7149-80-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty