Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-Amino-4-(3-chlorophenyl)butanoic acid 97%

Supplier: AMBEED, INC
Warning

A154826-100MG A154826-1G A154826-250MG
77137-902EA 73.13 USD InStock
77137-902 77137-904 77137-906
(S)-3-Amino-4-(3-chlorophenyl)butanoic acid 97%
(S)-3-Amino-4-(3-chlorophenyl)butanoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂ClNO₂
MDL Number: MFCD08461687
CAS Number: 270596-38-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty