Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorobenzhydrazide 95%

Supplier: AMBEED, INC
Warning

A156865-100G A156865-10G A156865-1G A156865-5G A156865-25G
77137-994EA 185.4 USD InStock
77137-994 77137-996 77137-998 77138-000 77746-074
2-Fluorobenzhydrazide 95%
2-Fluorobenzhydrazide
Precaución: for Research Use Only


Formula: C₇H₇FN₂O
MW: 154.14
MDL Number: MFCD00025112
CAS Number: 446-24-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty