Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-Methylsuccinic acid 98%

Supplier: AMBEED, INC
Warning

A159574-1G A159574-250MG A159574-25G A159574-5G
77138-080EA 33.99 USD InStock
77138-080 77138-082 77138-084 77138-086
(S)-(-)-Methylsuccinic acid 98%
(S)-(-)-Methylsuccinic acid
Precaución: for Research Use Only


Formula: C₅H₈O₄
MW: 132.115 g/mol
MDL Number: MFCD00192321
CAS Number: 2174-58-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty