Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyanophenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A163942-100G A163942-10G A163942-1G A163942-25G A163942-5G A163942-500G
77138-256EA 108.15 USD InStock
77138-256 77138-258 77138-260 77138-262 77138-264 77685-340
4-Cyanophenylboronic acid 98%
4-Cyanophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₆BNO₂
MDL Number: MFCD01318968
CAS Number: 126747-14-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty