Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Glu-OMe 98%

Supplier: AMBEED, INC
Warning

A169419-10G A169419-1G A169419-250MG A169419-25G A169419-5G
77138-376EA 29.87 USD InStock
77138-376 77138-378 77138-380 77138-382 77138-384
Fmoc-Glu-OMe 98%
Fmoc-Glu-OMe
Precaución: for Research Use Only


Formula: C₂₁H₂₁NO₆
MW: 383.39 g/mol
MDL Number: MFCD21607506
CAS Number: 145038-49-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty