Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-3759 98%

Supplier: AMBEED, INC
Warning

A171107-100MG A171107-10MG A171107-250MG A171107-50MG A171107-5MG A171107-5G
77138-406EA 21.63 USD InStock
77138-406 77138-408 77138-410 77138-412 77138-414 77685-381
AZD-3759 98%
AZD-3759
Precaución: for Research Use Only


Formula: C₂₂H₂₃ClFN₅O₃
MW: 459.9 g/mol
MDL Number: MFCD29058564
CAS Number: 1626387-80-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty