Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-((2-(methylthio)ethyl)amino)-2-((3,3,3-trifluoropropyl)thio)-9H-purin-9-yl)tetrahydrofuran-3,4-diol 97%

Supplier: AMBEED, INC
Warning

A174029-250MG A174029-1G
77138-508EA 14.42 USD InStock
77138-508 77685-389
(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-((2-(methylthio)ethyl)amino)-2-((3,3,3-trifluoropropyl)thio)-9H-purin-9-yl)tetrahydrofuran-3,4-diol 97%
(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-((2-(methylthio)ethyl)amino)-2-((3,3,3-trifluoropropyl)thio)-9H-purin-9-yl)tetrahydrofuran-3,4-diol
Precaución: for Research Use Only


Formula: C₁₆H₂₂F₃N₅O₄S₂
MW: 469.5 g/mol
MDL Number: MFCD15832982
CAS Number: 163706-58-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty