Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iminostilbene 97%

Supplier: AMBEED, INC
Warning

A175184-100G A175184-25G A175184-500G A175184-5G
77138-530EA 57.68 USD InStock
77138-530 77138-532 77138-534 77138-536
Iminostilbene 97%
Iminostilbene
Precaución: for Research Use Only


Formula: C₁₄H₁₁N
MW: 193.25 g/mol
MDL Number: MFCD00005071
CAS Number: 256-96-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty