Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorobenzamide 97%

Supplier: AMBEED, INC
Warning

A178481-100G A178481-25G A178481-500G A178481-5G
77138-628EA 51.51 USD InStock
77138-628 77138-630 77138-632 77138-634
4-Chlorobenzamide 97%
4-Chlorobenzamide
Precaución: for Research Use Only


Formula: C₇H₆ClNO
MDL Number: MFCD00007993
CAS Number: 619-56-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty