Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chlor-1,2-benzisothiazol 98%

Supplier: AMBEED, INC
Warning

A179929-100G A179929-10G A179929-25G
77138-678EA 31.93 USD InStock
77138-678 77138-680 77138-682
3-Chlor-1,2-benzisothiazol 98%
3-Chlor-1,2-benzisothiazol
Precaución: for Research Use Only


Formula: C₇H₄ClNS
MDL Number: MFCD00673254
CAS Number: 7716-66-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty