Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Terbinafine 98%

Supplier: AMBEED, INC
Warning

A180334-100MG A180334-10G A180334-1G A180334-250MG A180334-25G A180334-5G
77138-684EA 6.18 USD InStock
77138-684 77138-686 77138-688 77138-690 77138-692 77138-694
Terbinafine 98%
Terbinafine
Precaución: for Research Use Only


Formula: C₂₁H₂₅N
MW: 291.43 g/mol
MDL Number: MFCD00242672
CAS Number: 91161-71-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty