Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-fluoro-5-nitrotoluene 98%

Supplier: AMBEED, INC
Warning

A181145-10G A181145-1G A181145-250MG A181145-25G
77138-718EA 18.55 USD InStock
77138-718 77138-720 77138-722 77138-724
4-Chloro-2-fluoro-5-nitrotoluene 98%
4-Chloro-2-fluoro-5-nitrotoluene
Precaución: for Research Use Only


Formula: C₇H₅ClFNO₂
MDL Number: MFCD00134231
CAS Number: 18349-11-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty