Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-(trifluoromethoxy)benzene 98%

Supplier: AMBEED, INC
Danger

A182157-10G A182157-25G A182157-5G
77138-764EA 9.27 USD InStock
77138-764 77138-766 77138-768
1-Bromo-2-(trifluoromethoxy)benzene 98%
1-Bromo-2-(trifluoromethoxy)benzene
Precaución: for Research Use Only


Formula: C₇H₄BrF₃O
MDL Number: MFCD00040942
CAS Number: 64115-88-4
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty