Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzyloxy)propane-1,2-diol 97%

Supplier: AMBEED, INC
Warning

A183466-100G A183466-10G A183466-1G A183466-25G
77138-800EA 265.74 USD InStock
77138-800 77138-802 77138-804 77138-806
3-(Benzyloxy)propane-1,2-diol 97%
3-(Benzyloxy)propane-1,2-diol
Precaución: for Research Use Only


Formula: C₁₀H₁₄O₃
MW: 182.216 g/mol
MDL Number: MFCD00016633
CAS Number: 4799-67-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty