Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

I-BET726 98%

Supplier: AMBEED, INC
Warning

A186354-10MG A186354-1MG A186354-2MG A186354-5MG
77138-922EA 82.4 USD InStock
77138-922 77138-924 77138-926 77138-928
I-BET726 98%
I-BET726
Precaución: for Research Use Only


Formula: C₂₅H₂₃ClN₂O₃
MDL Number: MFCD28144683
CAS Number: 1300031-52-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty