Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nicotinamide 99+%

Warning

A187855-1000G A187855-100G A187855-25G A187855-500G A187855-5G
77138-964EA 42.44 USD InStock
77138-964 77138-966 77138-968 77138-970 77138-972
Nicotinamide 99+%
Nicotinamide
Precaución: for Research Use Only


Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 334
Flash Pt: 182
MDL Number: MFCD00006395
CAS Number: 98-92-0
Merck Index: 12,06574
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty