Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-1-naphthaleneboronic acid 97%

Supplier: AMBEED, INC
Warning

A191711-10G A191711-1G A191711-250MG A191711-25G A191711-5G
77139-058EA 28.84 USD InStock
77139-058 77139-060 77139-062 77139-064 77139-066
4-Methyl-1-naphthaleneboronic acid 97%
4-Methyl-1-naphthaleneboronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₁BO₂
MW: 186.02 g/mol
MDL Number: MFCD01632204
CAS Number: 103986-53-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty