Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-methylbenzenesulphonyl chloride 96%

Supplier: AMBEED, INC
Danger

A191948-10G A191948-1G A191948-250MG A191948-5G
77139-068EA 105.06 USD InStock
77139-068 77139-070 77139-072 77139-074
2-Fluoro-5-methylbenzenesulphonyl chloride 96%
2-Fluoro-5-methylbenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₇H₆ClFO₂S
MW: 208.64
MDL Number: MFCD04039785
CAS Number: 870704-14-6
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty