Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropyl (((((R)-1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)(phenoxy)phosphoryl)-L-alaninate hemifumarate 99+%

Supplier: AMBEED, INC
Warning

A192211-100MG A192211-10MG A192211-1MG A192211-250MG A192211-50MG A192211-5G A192211-5MG A192211-1G
77139-076EA 24.73 USD InStock
77139-076 77139-078 77139-080 77139-082 77139-084 77139-086 77139-088 77685-465
Isopropyl (((((R)-1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)(phenoxy)phosphoryl)-L-alaninate hemifumarate 99+%
Isopropyl (((((R)-1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)(phenoxy)phosphoryl)-L-alaninate hemifumarate
Precaución: for Research Use Only


Formula: C₄₆H₆₂N₁₂O₁₄P₂
MW: 1069.00 g/mol
MDL Number: MFCD30534429
CAS Number: 1392275-56-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty