Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromocumene 98%

Supplier: AMBEED, INC
Warning

A194070-100G A194070-10G A194070-1G A194070-25G A194070-500G A194070-5G
77139-166EA 101.97 USD InStock
77139-166 77139-168 77139-170 77139-172 77139-174 77139-176
2-Bromocumene 98%
2-Bromocumene
Precaución: for Research Use Only


Formula: C₉H₁₁Br
MDL Number: MFCD00051567
CAS Number: 7073-94-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty