Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Phenylcarbamoyl)benzeneboronic acid 98%

Supplier: AMBEED, INC
Warning

A195069-100MG A195069-1G A195069-250MG A195069-5G
77139-216EA 26.78 USD InStock
77139-216 77139-218 77139-220 77139-222
3-(Phenylcarbamoyl)benzeneboronic acid 98%
3-(Phenylcarbamoyl)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₂BNO₃
MDL Number: MFCD04115711
CAS Number: 397843-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty