Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Methyl-4-(trifluoromethyl)benzylamine 97%

Supplier: AMBEED, INC
Danger

A204729-1G A204729-250MG A204729-100MG
77139-448EA 46.35 USD InStock
77139-448 77139-450 77746-266
α-Methyl-4-(trifluoromethyl)benzylamine 97%
alpha-Methyl-4-(trifluoromethyl)benzylamine
Precaución: for Research Use Only


Formula: C₉H₁₀F₃N
MDL Number: MFCD04038878
CAS Number: 15996-84-6
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty