Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromoanthranilic acid 98%

Supplier: AMBEED, INC
Warning

A206280-100G A206280-25G A206280-5G
77139-508EA 118.45 USD InStock
77139-508 77139-510 77139-512
3,5-Dibromoanthranilic acid 98%
3,5-Dibromoanthranilic acid
Precaución: for Research Use Only


Formula: C₇H₅Br₂NO₂
MDL Number: MFCD00017087
CAS Number: 609-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty