Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1,5-Dimethyl-1H-pyrazol-3-yl)methanol 97%

Supplier: AMBEED, INC
Warning

A206628-10G A206628-1G A206628-250MG A206628-25G
77139-522EA 154.51 USD InStock
77139-522 77139-524 77139-526 77139-528
(1,5-Dimethyl-1H-pyrazol-3-yl)methanol 97%
(1,5-Dimethyl-1H-pyrazol-3-yl)methanol
Precaución: for Research Use Only


Formula: C₆H₁₀N₂O
MDL Number: MFCD03659702
CAS Number: 153912-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty