Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Divalproex sodium 98%

Supplier: AMBEED, INC
Warning

A206924-10MG A206924-1MG A206924-50MG A206924-100G A206924-1G
77139-534EA 9.26 USD InStock
77139-534 77139-536 77139-538 77685-535 77685-536
Divalproex sodium 98%
Divalproex sodium
Precaución: for Research Use Only


Formula: C₁₆H₃₁NaO₄
MDL Number: MFCD00879852
CAS Number: 76584-70-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty