Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrrolidin-1-ylacetonitrile 98%

Supplier: AMBEED, INC
Danger

A207148-10G A207148-1G A207148-25G A207148-5G
77139-540EA 53.56 USD InStock
77139-540 77139-542 77139-544 77139-546
Pyrrolidin-1-ylacetonitrile 98%
Pyrrolidin-1-ylacetonitrile
Precaución: for Research Use Only


Formula: C₆H₁₀N₂
MDL Number: MFCD00014098
CAS Number: 29134-29-0
UN: 3276
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty