Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fosinopril sodium 98%

Supplier: AMBEED, INC
Warning

A213166-10MG A213166-25MG A213166-50MG A213166-5MG
77139-658EA 7.21 USD InStock
77139-658 77139-660 77139-662 77139-664
Fosinopril sodium 98%
Fosinopril sodium
Precaución: for Research Use Only


Formula: C₃₀H₄₅NNaO₇P
MDL Number: MFCD23704184
CAS Number: 88889-14-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty