Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-m-xylene 97%

Supplier: AMBEED, INC
Warning

A214710-100G A214710-10G A214710-25G A214710-5G
77139-714EA 48.41 USD InStock
77139-714 77139-716 77139-718 77139-720
2-Chloro-m-xylene 97%
2-Chloro-m-xylene
Precaución: for Research Use Only


Formula: C₈H₉Cl
MDL Number: MFCD00000565
CAS Number: 6781-98-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty