Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-bromoisonicotinate 98%

Supplier: AMBEED, INC
Warning

A216439-10G A216439-1G A216439-250MG A216439-25G A216439-5G
77139-760EA 64.88 USD InStock
77139-760 77139-762 77139-764 77139-766 77139-768
Ethyl-2-bromoisonicotinate 98%
Ethyl-2-bromoisonicotinate
Precaución: for Research Use Only


Formula: C₈H₈BrNO₂
MW: 230.07 g/mol
MDL Number: MFCD06205252
CAS Number: 89978-52-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty