Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloroisoquinoline-1-carbonitrile 97%

Supplier: AMBEED, INC
Warning

A219105-100MG A219105-1G A219105-250MG
77139-822EA 13.39 USD InStock
77139-822 77139-824 77139-826
5-Chloroisoquinoline-1-carbonitrile 97%
5-Chloroisoquinoline-1-carbonitrile
Precaución: for Research Use Only


Formula: C₁₀H₅ClN₂
MDL Number: MFCD22096567
CAS Number: 1231761-25-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty