Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-methyl-6-nitroaniline 98%

Supplier: AMBEED, INC
Warning

A225334-100G A225334-10G A225334-1G A225334-25G A225334-5G
77139-918EA 73.13 USD InStock
77139-918 77139-920 77139-922 77139-924 77139-926
4-Bromo-2-methyl-6-nitroaniline 98%
4-Bromo-2-methyl-6-nitroaniline
Precaución: for Research Use Only


Formula: C₇H₇BrN₂O₂
MW: 231.05
Melting Pt: 155
MDL Number: MFCD00052919
CAS Number: 77811-44-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty