Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,5-Diphenylhydantoin sodium salt 98%

Supplier: AMBEED, INC
Danger

A228712-25G A228712-5G
77140-000EA 12.36 USD InStock
77140-000 77140-002
5,5-Diphenylhydantoin sodium salt 98%
5,5-Diphenylhydantoin sodium salt
Precaución: for Research Use Only


Formula: C₁₅H₁₂N₂O₂
MW: 274.25 g/mol
MDL Number: MFCD00069674
CAS Number: 630-93-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty